Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cdh4
Entrez Gene ID
12561
Gene Name
cadherin 4
Gene Aliases
AW120700, R-Cadh, R-cadherin, Rcad
Chromosome Location
Chr.2: 179442478 - 179899377 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009867.2 NP_033997.1 13 - 14 2378 60
GenBank mRNA D14888.1 - 13 - 14 2288 60
X69966.1 - 13 - 14 2234 60

View Full Product Details