Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Letm2
Entrez Gene ID
270035
Gene Name
leucine zipper-EF-hand containing transmembrane protein 2
Gene Aliases
6030453H13, D030041N04Rik
Chromosome Location
Chr.8: 25578481 - 25597899 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_173012.3 NP_766600.2 2 - 3 459 122
GenBank mRNA AK031563.1 - 2 - 3 313 122
AK034050.1 - 2 - 3 470 122
AK050943.1 - 2 - 3 625 122
AK163507.1 - 2 - 3 314 122
BC055685.1 - 2 - 3 459 122

View Full Product Details