The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011960.2 | NP_036090.2 | 17 - 18 | 3027 | 99 |
GenBank mRNA | AF079557.1 | - | 17 - 18 | 2776 | 99 |
AK034542.1 | - | 10 - 11 | 959 | 99 | |
AK088708.1 | - | 14 - 15 | 1382 | 99 | |
AK139114.1 | - | 2 - 3 | 242 | 99 | |
AK164013.1 | - | 17 - 18 | 3015 | 99 | |
BC050892.1 | - | 17 - 18 | 3026 | 99 | |
BC053037.1 | - | 17 - 18 | 3011 | 99 | |
BC059827.1 | - | 17 - 18 | 3036 | 99 | |
EF382673.1 | - | 15 - 16 | 1736 | 99 | |
JQ890227.1 | - | 14 - 15 | 1533 | 99 |