Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Drc1
Entrez Gene ID
381738
Gene Name
dynein regulatory complex subunit 1
Gene Aliases
Ccdc164, Gm1060, b2b1654Clo
Chromosome Location
Chr.5: 30341663 - 30366708 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033460.3 NP_001028632.1 - 743 57
GenBank mRNA AK140154.1 - - 744 57
BC150863.1 - - 741 57

View Full Product Details