The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001167879.1 | NP_001161351.1 | 4 - 5 | 1537 | 105 |
GenBank mRNA | BC060268.1 | - | 3 - 4 | 1321 | 105 |