Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ube3c
Entrez Gene ID
100763
Gene Name
ubiquitin protein ligase E3C
Gene Aliases
AI853514, mKIAA0010
Chromosome Location
Chr.5: 29569242 - 29676077 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_133907.3 NP_598668.1 2 - 3 397 141
GenBank mRNA AK029973.1 - 2 - 3 207 141
AK045211.1 - 2 - 3 194 141
AK046056.1 - 2 - 3 398 141
AK122187.1 - 2 - 3 332 141
AK140491.1 - 2 - 3 154 141
BC120731.1 - 2 - 3 144 141
BC137626.1 - 2 - 3 144 141

View Full Product Details