The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001293703.1 | NP_001280632.1 | 3 - 4 | 305 | 95 |
NM_028000.1 | NP_082276.1 | 3 - 4 | 305 | 95 | |
NR_121642.1 | 3 - 4 | 265 | 95 | ||
NR_121643.1 | 3 - 4 | 265 | 95 | ||
GenBank mRNA | AA184457.1 | - | 3 - 4 | 305 | 95 |
AK007542.1 | - | 1 - 2 | 266 | 95 | |
AK009455.1 | - | 3 - 4 | 272 | 95 | |
AK144593.1 | - | 3 - 4 | 290 | 95 | |
BC039626.1 | - | 2 - 3 | 120 | 95 | |
BC062173.1 | - | 3 - 4 | 238 | 95 | |
BC089556.1 | - | 3 - 4 | 249 | 95 | |
BC099489.1 | - | 3 - 4 | 256 | 95 |