The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_178390.3 | NP_848477.2 | 5 - 6 | 1473 | 75 |
GenBank mRNA | AK010537.1 | - | 5 - 6 | 1409 | 75 |
AK017321.1 | - | 4 - 5 | 1818 | 75 | |
AK155077.1 | - | 6 - 7 | 1586 | 75 | |
AK163326.1 | - | 5 - 6 | 1445 | 75 | |
BC016596.1 | - | 3 - 4 | 898 | 75 |