Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Parg
Entrez Gene ID
26430
Gene Name
poly (ADP-ribose) glycohydrolase
Gene Aliases
AI413217
Chromosome Location
Chr.14: 32201949 - 32298714 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK080306.1 - 1 - 2 213 85

View Full Product Details