Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Insr
Entrez Gene ID
16337
Gene Name
insulin receptor
Gene Aliases
4932439J01Rik, CD220, D630014A15Rik, IR, IR-A, IR-B
Chromosome Location
Chr.8: 3150922 - 3279649 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_010568.2 NP_034698.2 20 - 21 4256 84
GenBank mRNA AK164267.1 - 2 - 3 252 84
J05149.1 - 20 - 21 3815 84
KC356871.1 - 21 - 22 3803 84

View Full Product Details