Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Insr
Entrez Gene ID
16337
Gene Name
insulin receptor
Gene Aliases
4932439J01Rik, CD220, D630014A15Rik, IR, IR-A, IR-B
Chromosome Location
Chr.8: 3150922 - 3279649 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_010568.2 NP_034698.2 9 - 10 2524 108
GenBank mRNA EU346716.1 - 2 - 3 340 108
J05149.1 - 9 - 10 2083 108
KC356871.1 - 9 - 10 2035 108

View Full Product Details