Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nfxl1
Entrez Gene ID
100978
Gene Name
nuclear transcription factor, X-box binding-like 1
Gene Aliases
1700012H24Rik, AW538212, D430033A06Rik, DNABF, GCF, Gm1058, TCF9
Chromosome Location
Chr.5: 72513301 - 72559707 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_133921.2 NP_598682.2 11 - 12 1532 77
GenBank mRNA DV657577.1 - 5 - 6 483 77

View Full Product Details