The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_017471.2 | NP_059499.1 | 6 - 7 | 562 | 109 |
GenBank mRNA | AF005204.1 | - | 6 - 7 | 566 | 109 |
AK137501.1 | - | 6 - 7 | 589 | 109 |