Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Txk
Entrez Gene ID
22165
Gene Name
TXK tyrosine kinase
Gene Aliases
A130089B16Rik, Btkl, PTK-RL-18, PTK4, Rlk
Chromosome Location
Chr.5: 72695978 - 72752992 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001122754.2 NP_001116226.1 7 - 8 657 125
NM_001289494.1 NP_001276423.1 6 - 7 591 125
NM_001289495.1 NP_001276424.1 7 - 8 674 125
NM_013698.3 NP_038726.2 6 - 7 741 125
GenBank mRNA AK088209.1 - 6 - 7 587 125
AK138069.1 - 7 - 8 654 125
AK138786.1 - 7 - 8 125
BC120552.1 - 6 - 7 644 125
BC137710.1 - 6 - 7 646 125
D43963.1 - 6 - 7 741 125
D83201.1 - 6 - 7 681 125
D83202.1 - 6 - 7 844 125
U16145.1 - 7 - 8 622 125
U19607.1 - 7 - 8 657 125

View Full Product Details