The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_026660.2 | NP_080936.1 | 12 - 13 | 1454 | 118 |
GenBank mRNA | AK003763.1 | - | 7 - 8 | 722 | 118 |
AK018736.1 | - | 12 - 13 | 1467 | 118 | |
AK089055.1 | - | 12 - 13 | 1471 | 118 | |
AK144407.1 | - | 3 - 4 | 174 | 118 | |
AK152438.1 | - | 12 - 13 | 1440 | 118 | |
BC023012.1 | - | 12 - 13 | 1454 | 118 | |
BC064775.1 | - | 12 - 13 | 1352 | 118 |