The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_022656.2 | NP_073147.2 | 20 - 21 | 4337 | 80 |
GenBank mRNA | AF315344.1 | - | 16 - 17 | 3790 | 80 |
AK122412.1 | - | 18 - 19 | 3835 | 80 | |
AK191277.1 | - | 1 - 2 | 140 | 80 | |
AK195831.1 | - | 1 - 2 | 140 | 80 | |
AY032852.1 | - | 3 - 4 | 472 | 80 | |
BC089329.1 | - | 15 - 16 | 3556 | 80 | |
BC108364.1 | - | 15 - 16 | 3532 | 80 | |
BC144980.1 | - | 16 - 17 | 3666 | 80 |