The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001165930.1 | NP_001159402.1 | 9 - 10 | 994 | 102 |
NM_029626.2 | NP_083902.2 | 9 - 10 | 1523 | 102 | |
GenBank mRNA | AK010388.1 | - | 9 - 10 | 1252 | 102 |
AK077388.1 | - | 9 - 10 | 987 | 102 | |
BC029215.1 | - | 7 - 8 | 1195 | 102 | |
BC055709.1 | - | 8 - 9 | 1410 | 102 | |
BC058731.1 | - | 8 - 9 | 1410 | 102 | |
BC069873.1 | - | 9 - 10 | 1073 | 102 |