Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Erlin1
Entrez Gene ID
226144
Gene Name
ER lipid raft associated 1
Gene Aliases
2810439N09Rik, C80197, Keo4, Spfh1
Chromosome Location
Chr.19: 44034943 - 44070493 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001164360.1 NP_001157832.1 1 - 2 96 74
GenBank mRNA AK145990.1 - 1 - 2 69 74
BC011220.1 - 1 - 2 60 74
CJ074662.1 - 1 - 2 97 74

View Full Product Details