Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Erlin1
Entrez Gene ID
226144
Gene Name
ER lipid raft associated 1
Gene Aliases
2810439N09Rik, C80197, Keo4, Spfh1
Chromosome Location
Chr.19: 44034943 - 44070493 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001164359.1 NP_001157831.1 6 - 7 625 120
NM_001164360.1 NP_001157832.1 6 - 7 533 120
NM_145502.3 NP_663477.3 5 - 6 729 120
GenBank mRNA AK133557.1 - 5 - 6 472 120
AK145990.1 - 6 - 7 506 120
BC011220.1 - 6 - 7 497 120
BC023849.1 - 5 - 6 592 120

View Full Product Details