Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Erg
- Entrez Gene ID
-
13876
- Gene Name
- avian erythroblastosis virus E-26 (v-ets) oncogene related
- Gene Aliases
-
D030036I24Rik
- Chromosome Location
-
Chr.16: 95359169 - 95586593 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BB639043.1 |
- |
1 - 2 |
123 |
141 |
View Full Product Details