The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015775.2 | NP_056590.2 | 5 - 6 | 675 | 65 |
GenBank mRNA | AF113596.1 | - | 5 - 6 | 545 | 65 |
AF199362.1 | - | 5 - 6 | 556 | 65 | |
AF243500.1 | - | 5 - 6 | 545 | 65 | |
AK141350.1 | - | 5 - 6 | 650 | 65 | |
AK146047.1 | - | 5 - 6 | 559 | 65 | |
AK161173.1 | - | 5 - 6 | 669 | 65 | |
BC038393.1 | - | 5 - 6 | 514 | 65 | |
BC054348.1 | - | 5 - 6 | 675 | 65 |