Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Nudt22
Entrez Gene ID
68323
Gene Name
nudix (nucleoside diphosphate linked moiety X)-type motif 22
Gene Aliases
0610006K04Rik, AI851793, AW545547
Chromosome Location
Chr.19: 6993019 - 6996115 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_026675.2 NP_080951.1 3 - 4 620 88
GenBank mRNA AK002252.1 - 3 - 4 621 88
BC019768.1 - 2 - 3 597 88

View Full Product Details