The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001127355.1 | NP_001120827.1 | 11 - 12 | 1447 | 76 |
NM_001127356.1 | NP_001120828.1 | 10 - 11 | 1669 | 76 | |
NM_010122.2 | NP_034252.2 | 11 - 12 | 1302 | 76 | |
GenBank mRNA | AK154364.1 | - | 11 - 12 | 1210 | 76 |
AK158282.1 | - | 11 - 12 | 1211 | 76 | |
BC021884.1 | - | 11 - 12 | 1189 | 76 | |
CD768292.1 | - | 2 - 3 | 197 | 76 | |
M98035.1 | - | 10 - 11 | 1289 | 76 | |
M98036.1 | - | 11 - 12 | 1255 | 76 |