Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Nsg1
Entrez Gene ID
18196
Gene Name
neuron specific gene family member 1
Gene Aliases
Neep21, m234, p21
Chromosome Location
Chr.5: 38137192 - 38159467 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_010942.3 NP_035072.2 2 - 3 232 100
GenBank mRNA AF035683.1 - 2 - 3 181 100
AK076451.1 - 2 - 3 672 100
AK078054.1 - 2 - 3 223 100
AK144503.1 - 2 - 3 207 100
AK148941.1 - 2 - 3 232 100
AX775765.1 - 2 - 3 232 100
BC008272.1 - 2 - 3 218 100

View Full Product Details