Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ddx58
Entrez Gene ID
230073
Gene Name
DEAD (Asp-Glu-Ala-Asp) box polypeptide 58
Gene Aliases
6430573D20Rik, C330021E21, RIG-I, RLR-1
Chromosome Location
Chr.4: 40203777 - 40239825 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_172689.3 NP_766277.3 16 - 17 2470 83
GenBank mRNA AK049305.1 - 7 - 8 1061 83
AK128929.1 - 16 - 17 2405 83
AK155930.1 - 15 - 16 3362 83
BC127163.1 - 16 - 17 2336 83
BC152540.1 - 16 - 17 2338 83

View Full Product Details