The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001098230.1 | NP_001091700.1 | 2 - 3 | 187 | 69 |
GenBank mRNA | BB611000.1 | - | 2 - 3 | 160 | 69 |
CJ147172.1 | - | 2 - 3 | 165 | 69 |