The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_144541.1 | NP_653124.1 | 10 - 11 | 985 | 74 |
NM_181279.1 | NP_851796.1 | 12 - 13 | 1096 | 74 | |
NM_181280.1 | NP_851797.1 | 11 - 12 | 994 | 74 | |
NM_181281.1 | NP_851798.1 | 9 - 10 | 829 | 74 | |
NM_181282.1 | NP_851799.1 | 10 - 11 | 906 | 74 | |
GenBank mRNA | AF440752.1 | - | 10 - 11 | 985 | 74 |
AF527952.1 | - | 12 - 13 | 1096 | 74 | |
AF527953.1 | - | 10 - 11 | 906 | 74 | |
AF527954.1 | - | 10 - 11 | 916 | 74 | |
AF527955.1 | - | 9 - 10 | 829 | 74 | |
AF538925.1 | - | 11 - 12 | 994 | 74 | |
AK080991.1 | - | 10 - 11 | 1021 | 74 | |
AK156929.1 | - | 10 - 11 | 1077 | 74 | |
BC061000.1 | - | 10 - 11 | 1061 | 74 | |
BC100565.1 | - | 6 - 7 | 540 | 74 |