Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mcph1
Entrez Gene ID
244329
Gene Name
microcephaly, primary autosomal recessive 1
Gene Aliases
5430437K10Rik, BRIT1, D030046N04Rik, MCT
Chromosome Location
Chr.8: 18595136 - 18803189 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_173189.2 NP_775281.2 6 - 7 999 72
GenBank mRNA AK042026.1 - 5 - 6 893 72
AK083572.1 - 6 - 7 1042 72
AK147997.1 - 6 - 7 591 72
AK148021.1 - 6 - 7 597 72
AY070216.1 - 6 - 7 579 72
BC056924.1 - 6 - 7 999 72

View Full Product Details