Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pax2
Entrez Gene ID
18504
Gene Name
paired box 2
Gene Aliases
Opdc, Pax-2
Chromosome Location
Chr.19: 44745695 - 44837871 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011037.4 NP_035167.4 9 - 10 1103 99
GenBank mRNA BC148232.1 - 9 - 10 1107 99
BC150484.1 - 9 - 10 1107 99

View Full Product Details