The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_028102.1 | NP_082378.1 | 2 - 3 | 324 | 66 |
GenBank mRNA | AK008529.1 | - | 2 - 3 | 310 | 66 |
AK035532.1 | - | 2 - 3 | 376 | 66 | |
AK079311.1 | - | 2 - 3 | 372 | 66 | |
AK173010.1 | - | 2 - 3 | 276 | 66 | |
AK196094.1 | - | 1 - 2 | 240 | 66 | |
BC046229.1 | - | 1 - 2 | 244 | 66 | |
BC118962.2 | - | 2 - 3 | 559 | 66 |