Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Plekha2
Entrez Gene ID
83436
Gene Name
pleckstrin homology domain-containing, family A (phosphoinositide binding specific) member 2
Gene Aliases
6430512N22Rik, AI851530, AU041791, TAPP2, W91773
Chromosome Location
Chr.8: 25039144 - 25104917 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_031257.3 NP_112547.1 10 - 11 1014 61
GenBank mRNA AF286161.1 - 10 - 11 968 61
AF418551.1 - 9 - 10 880 61
AK032325.1 - 10 - 11 1023 61
AK042324.1 - 10 - 11 988 61
AK078212.1 - 10 - 11 993 61
AK088395.1 - 10 - 11 956 61
AK153117.1 - 10 - 11 986 61
BC010215.1 - 10 - 11 935 61

View Full Product Details