Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Fcrl1
- Entrez Gene ID
-
229499
- Gene Name
- Fc receptor-like 1
- Gene Aliases
-
A230020G22Rik, Bxmas1, Bxmh1, Bxmh1b, Fcrh1, Fcrh1l, Fcrh1s, Ifgp1
- Chromosome Location
-
Chr.3: 87376333 - 87402934 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC055830.1 |
- |
10 - 11 |
1033 |
98 |
View Full Product Details