The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_133903.3 | NP_598664.3 | 1 - 2 | 288 | 54 |
GenBank mRNA | AK028987.1 | - | 1 - 2 | 233 | 54 |
AK042840.1 | - | 1 - 2 | 236 | 54 | |
BB848417.1 | - | 1 - 2 | 288 | 54 |