Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Wdr95
Entrez Gene ID
381693
Gene Name
WD40 repeat domain 95
Gene Aliases
4930434E21Rik
Chromosome Location
Chr.5: 149528679 - 149611894 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_029440.3 NP_083716.2 20 - 21 1790 102
GenBank mRNA AK015309.1 - 18 - 19 1894 102
AK087389.1 - 20 - 21 1790 102

View Full Product Details