Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Uspl1
Entrez Gene ID
231915
Gene Name
ubiquitin specific peptidase like 1
Gene Aliases
E430026A01Rik
Chromosome Location
Chr.5: 149184560 - 149215434 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001013378.2 NP_001013396.2 5 - 6 1258 162
NM_001115149.1 NP_001108621.1 5 - 6 1258 162
NM_001115150.1 NP_001108622.1 4 - 5 1089 162
NM_001115151.1 NP_001108623.1 4 - 5 1089 162
NM_001115153.1 NP_001108625.1 6 - 7 1435 162
NM_001286824.1 NP_001273753.1 5 - 6 1216 162
NM_001286825.1 NP_001273754.1 5 - 6 1200 162
GenBank mRNA AK050969.1 - 5 - 6 1242 162
AK088787.1 - 4 - 5 955 162
AK145409.1 - 6 - 7 1333 162
AK146820.1 - 5 - 6 1083 162
AK161654.1 - 4 - 5 936 162
AK168702.1 - 5 - 6 1083 162
AK168981.1 - 5 - 6 1110 162
BC028849.1 - 5 - 6 1152 162
BC032291.1 - 2 - 3 3228 162
BC086664.1 - 5 - 6 1100 162

View Full Product Details