Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Uspl1
Entrez Gene ID
231915
Gene Name
ubiquitin specific peptidase like 1
Gene Aliases
E430026A01Rik
Chromosome Location
Chr.5: 149184560 - 149215434 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001013378.2 NP_001013396.2 9 - 10 1843 62
NM_001115151.1 NP_001108623.1 8 - 9 1674 62
GenBank mRNA AK161654.1 - 8 - 9 1521 62
BC032291.1 - 6 - 7 3813 62
BC086664.1 - 9 - 10 1685 62

View Full Product Details