Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cbwd1
Entrez Gene ID
226043
Gene Name
COBW domain containing 1
Gene Aliases
AV349248, BC018472
Chromosome Location
Chr.19: 24919916 - 24961616 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_146097.3 NP_666209.1 9 - 10 785 193
NR_033744.1 9 - 10 852 193
GenBank mRNA AK132291.1 - 9 - 10 771 193
BC018472.1 - 9 - 10 727 193
BC053745.1 - 9 - 10 820 193

View Full Product Details