Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mob3b
Entrez Gene ID
214944
Gene Name
MOB kinase activator 3B
Gene Aliases
8430436F23Rik, A430018A01Rik, Mobkl2b
Chromosome Location
Chr.4: 34949074 - 35157495 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_178061.5 NP_835162.1 1 - 2 238 58
GenBank mRNA AK039841.1 - 1 - 2 84 58
AK087285.1 - 1 - 2 239 58
AK155217.1 - 1 - 2 239 58
AK170973.1 - 1 - 2 232 58
BC020028.1 - 1 - 2 220 58

View Full Product Details