Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- LOC105247168
- Entrez Gene ID
-
105247168
- Gene Name
- uncharacterized LOC105247168
- Gene Aliases
-
-
- Chromosome Location
-
Chr.4: 35225204 - 35235851 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK045932.1 |
- |
- |
255 |
78 |
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
BB644480.1 |
- |
1 - 2 |
255 |
|
Chr. 4 - 35225932 - 35260732 [+] on Build GRCm38 |
78 |
Adjacent Gene Details
|
Gene Name |
Gene Symbol |
Alias |
Distance |
Relative Direction |
Location Chromosome |
Upstream |
MOB kinase activator 3B |
Mob3b |
8430436F23Rik, A430018A01Rik, Mobkl2b |
-68.4kb
|
Anti-sense |
Chr. - 34949074 - 35157495 on Build GRCm38 |
Downstream |
leucine rich repeat and Ig domain containing 2 |
Lingo2 |
B230217C06Rik, Lern3, Lrrn6c |
445.9kb
|
Anti-sense |
Chr. - 35706647 - 36952254 on Build GRCm38 |
View Full Product Details