The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001271725.1 | NP_001258654.1 | 10 - 11 | 2137 | 112 |
NM_001271726.1 | NP_001258655.1 | 10 - 11 | 2173 | 112 | |
NM_001271727.1 | NP_001258656.1 | 10 - 11 | 2442 | 112 | |
NM_001271728.1 | NP_001258657.1 | 10 - 11 | 2240 | 112 | |
NM_030706.3 | NP_109631.2 | 10 - 11 | 2236 | 112 | |
GenBank mRNA | AB043550.1 | - | 10 - 11 | 2220 | 112 |
AF220017.1 | - | 10 - 11 | 2085 | 112 | |
AK042752.1 | - | 2 - 3 | 229 | 112 | |
AK045410.1 | - | 10 - 11 | 2233 | 112 | |
AK079415.1 | - | 10 - 11 | 2387 | 112 | |
AK141419.1 | - | 10 - 11 | 2083 | 112 | |
AK147275.1 | - | 10 - 11 | 2350 | 112 | |
AK147395.1 | - | 10 - 11 | 2231 | 112 | |
AK164350.1 | - | 9 - 10 | 1939 | 112 | |
AK220343.1 | - | 9 - 10 | 3679 | 112 | |
BC058668.1 | - | 9 - 10 | 3048 | 112 | |
BC058961.1 | - | 10 - 11 | 2240 | 112 |