The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001045529.3 | NP_001038994.2 | 7 - 8 | 1072 | 126 |
GenBank mRNA | AK132226.1 | - | 7 - 8 | 928 | 126 |
AK144885.1 | - | 8 - 9 | 1027 | 126 | |
AK172896.1 | - | 7 - 8 | 1072 | 126 | |
BC098072.1 | - | 7 - 8 | 913 | 126 | |
BC145705.1 | - | 7 - 8 | 1021 | 126 |