The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_028150.2 | NP_082426.1 | 1 - 2 | 150 | 108 |
GenBank mRNA | AK012134.1 | - | 1 - 2 | 96 | 108 |
AK047063.1 | - | 1 - 2 | 120 | 108 | |
BC031447.1 | - | 1 - 2 | 73 | 108 | |
BY129485.1 | - | 1 - 2 | 151 | 108 |