The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016807.2 | NP_058087.2 | 4 - 5 | 341 | 82 |
GenBank mRNA | AF077527.1 | - | 4 - 5 | 300 | 82 |
AK150661.1 | - | 4 - 5 | 309 | 82 | |
AK150736.1 | - | 4 - 5 | 309 | 82 | |
AK159323.1 | - | 4 - 5 | 287 | 82 | |
AK159538.1 | - | 4 - 5 | 314 | 82 | |
AK165657.1 | - | 4 - 5 | 311 | 82 |