The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009524.3 | NP_033550.2 | 1 - 2 | 647 | 77 |
GenBank mRNA | AK031512.1 | - | 1 - 2 | 357 | 77 |
AK031597.1 | - | 1 - 2 | 620 | 77 | |
BC018425.1 | - | 1 - 2 | 264 | 77 | |
M89798.1 | - | 1 - 2 | 561 | 77 |