Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tubgcp3
Entrez Gene ID
259279
Gene Name
tubulin, gamma complex associated protein 3
Gene Aliases
GCP3, Spc98p
Chromosome Location
Chr.8: 12614277 - 12672243 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_198031.1 NP_932148.1 6 - 7 781 80
GenBank mRNA AK051775.1 - 6 - 7 793 80
AK150149.1 - 6 - 7 805 80
AK165503.1 - 6 - 7 804 80
AK171491.1 - 6 - 7 787 80
AK171804.1 - 6 - 7 796 80
BC046615.1 - 6 - 7 781 80
BC058566.1 - 6 - 7 781 80

View Full Product Details