Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sdr16c6
Entrez Gene ID
242286
Gene Name
short chain dehydrogenase/reductase family 16C, member 6
Gene Aliases
4833413O15Rik, Gm423
Chromosome Location
Chr.4: 4055926 - 4077594 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001080710.2 NP_001074179.1 6 - 7 947 86
GenBank mRNA BC125450.1 - 6 - 7 947 86
BC125452.1 - 6 - 7 947 86

View Full Product Details