Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tmem47
Entrez Gene ID
192216
Gene Name
transmembrane protein 47
Gene Aliases
3010015F07Rik, AI115536, BCMP1, Tm4sf10
Chromosome Location
Chr.X: 81070644 - 81097875 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_138751.2 NP_620090.1 2 - 3 658 106
GenBank mRNA AB041540.1 - 2 - 3 591 106
AK030761.1 - 2 - 3 587 106
AK076147.1 - 2 - 3 597 106
AK133790.1 - 2 - 3 595 106
AK162304.1 - 2 - 3 596 106
BC019751.1 - 2 - 3 569 106

View Full Product Details