The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001159774.1 | NP_001153246.1 | 4 - 5 | 452 | 63 |
NM_010546.2 | NP_034676.1 | 4 - 5 | 452 | 63 | |
GenBank mRNA | AF026524.1 | - | 3 - 4 | 320 | 63 |
AF088910.1 | - | 4 - 5 | 512 | 63 | |
AK011359.1 | - | 4 - 5 | 419 | 63 | |
AK086442.1 | - | 4 - 5 | 448 | 63 | |
AK139972.1 | - | 4 - 5 | 410 | 63 | |
AK144048.1 | - | 4 - 5 | 429 | 63 | |
AK144603.1 | - | 4 - 5 | 445 | 63 | |
AK150884.1 | - | 4 - 5 | 404 | 63 | |
AK151888.1 | - | 4 - 5 | 404 | 63 | |
AK151955.1 | - | 4 - 5 | 404 | 63 | |
AK153151.1 | - | 4 - 5 | 404 | 63 | |
AK155587.1 | - | 4 - 5 | 404 | 63 | |
AK156294.1 | - | 4 - 5 | 410 | 63 | |
AK182153.1 | - | 1 - 2 | 92 | 63 | |
AK201390.1 | - | 2 - 3 | 230 | 63 | |
BC004772.1 | - | 4 - 5 | 390 | 63 | |
BC037723.1 | - | 4 - 5 | 424 | 63 |