The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001113362.1 | NP_001106833.1 | 4 - 5 | 1082 | 66 |
NM_133910.3 | NP_598671.3 | 3 - 4 | 901 | 66 | |
GenBank mRNA | AB093293.1 | - | 2 - 3 | 695 | 66 |
AK029986.1 | - | 4 - 5 | 1083 | 66 | |
BC042515.1 | - | 3 - 4 | 877 | 66 | |
BC086316.1 | - | 3 - 4 | 969 | 66 | |
BC096446.1 | - | 3 - 4 | 893 | 66 |