Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Paxip1
Entrez Gene ID
55982
Gene Name
PAX interacting (with transcription-activation domain) protein 1
Gene Aliases
D5Ertd149e, PTIP
Chromosome Location
Chr.5: 27740080 - 27792079 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018878.3 NP_061366.2 20 - 21 3417 69
GenBank mRNA AF104261.1 - 20 - 21 3417 69
AK144606.1 - 20 - 21 3279 69
AK197074.1 - 1 - 2 40 69
BC066014.1 - 20 - 21 3206 69

View Full Product Details